Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H40009

Sigma-Aldrich

2-Hydroxy-2-methylbutyric acid

98%

Synonym(s):

α-Hydroxy-α-methylbutyric acid, (±)-α-Hydroxy-α-methylbutyric acid, (±)-2-Hydroxy-2-methylbutanoic acid, (±)-2-Hydroxy-2-methylbutyric acid, 2-Hydroxy-2-methylbutanoic acid, 2-Methyl-2-hydroxybutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C(CH3)(OH)CO2H
CAS Number:
Molecular Weight:
118.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

98%

mp

73-75 °C (lit.)

SMILES string

CCC(C)(O)C(O)=O

InChI

1S/C5H10O3/c1-3-5(2,8)4(6)7/h8H,3H2,1-2H3,(H,6,7)

InChI key

MBIQENSCDNJOIY-UHFFFAOYSA-N

Application

2-Hydroxy-2-methylbutyric acid can be used as:
  • A complexing agent in a new electrolytic system, applicable in the isotachophoretic lanthanide separation.[1]
  • An aldehyde surrogate to prepare pyrrolo[1,2-a]quinoxaline derivatives by reacting with 2-(1H-pyrrol-1-yl)aniline.[2]
  • A starting material to synthesize Cr(V) reagent named sodium bis(2-hydroxy-2-methylbutyrato)oxochromate(V) (Aldrich cat. no. ALD00006) applicable in the total synthesis of (−)-taxuyunnanine D.[3]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Separation and analysis of lanthanides by isotachophoresis coupled with inductively coupled plasma mass spectrometry
Vio L, et al.
Talanta, 99, 586-593 (2012)
Two-phase synthesis of (−)-taxuyunnanine D
Wilde NC, et al.
Journal of the American Chemical Society, 136(13), 4909-4912 (2014)
Florence Guéguen et al.
Talanta, 162, 278-284 (2016-11-14)
The high-precision isotopic characterization of actinides and fission products in nuclear samples is fundamental for various applications such as the management of spent nuclear fuel or the validation of neutronic calculation codes. However multi-elemental isotope ratio measurements by mass spectrometric
W Dekant et al.
Research report (Health Effects Institute), (102)(102), 29-71 (2001-08-16)
The biotransformation of methyl tert-butyl ether (MTBE), ethyl tert-butyl ether (ETBE), and tert-amyl methyl ether (TAME) was studied in humans and in rats after inhalation of 4 and 40 ppm of MTBE, ETBE, and TAME, respectively, for 4 hours, and
α-Hydroxy acid as an aldehyde surrogate: metal-free synthesis of pyrrolo [1, 2-a] quinoxalines, quinazolinones, and other N-heterocycles via decarboxylative oxidative annulation reaction
Viji M, et al.
Royal Society of Chemistry Advances, 10(61), 37202-37208 (2020)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service