Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF97526784

Nickel-chromium alloy, Ni80Cr20

foil, 25 x 25mm, 0.005mm thickness, as rolled, not light tested (nlt)

Synonym(s):

NI050200

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101711
NACRES:
NA.23

description

not light tested (nlt)

form

foil (as rolled)

manufacturer/tradename

Goodfellow 975-267-84

size × thickness

25 mm × 25 mm × 0.005 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service