Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

GF26982557

Vanadium

foil, thickness 1.0 mm, size 25 × 25 mm, purity 99.8%

Synonym(s):

V 000340

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
V
CAS Number:
Molecular Weight:
50.94
MDL number:
UNSPSC Code:
12141749
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

8 mmHg ( 20 °C)

assay

99.8%

form

foil

manufacturer/tradename

Goodfellow 269-825-57

resistivity

24.8-26.0 μΩ-cm, 20°C

size

25 × 25 mm

thickness

1.0 mm

bp

3380 °C (lit.)

mp

1890 °C (lit.)

density

6.11 g/mL at 25 °C (lit.)

SMILES string

[V]

InChI

1S/V

InChI key

LEONUFNNVUYDNQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service