Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D89905

Sigma-Aldrich

N,N-Diethylaniline

98%

Synonym(s):

DEA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NC6H5
CAS Number:
Molecular Weight:
149.23
Beilstein/REAXYS Number:
742483
EC Number:
MDL number:

vapor density

5.2 (vs air)

vapor pressure

1 mmHg ( 49.7 °C)

assay

98%

autoignition temp.

1166 °F

refractive index

n20/D 1.542 (lit.)

bp

217 °C (lit.)

mp

−38 °C (lit.)

density

0.938 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)c1ccccc1

InChI

1S/C10H15N/c1-3-11(4-2)10-8-6-5-7-9-10/h5-9H,3-4H2,1-2H3

InChI key

GGSUCNLOZRCGPQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

DuPont product

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Skin Irrit. 2 - STOT RE 2 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 2

flash_point_f

190.4 °F - closed cup

flash_point_c

88 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giorgio Ottaviani et al.
Journal of medicinal chemistry, 50(4), 742-748 (2007-02-16)
The development of in silico and in vitro tools to estimate or predict the passive human skin permeation and distribution of new chemical entities, useful in dermal drug delivery, in absorption studies of toxic compounds, and in the cosmetics industry

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service