Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D75408

Sigma-Aldrich

1,3-Dichlorotetrafluoroacetone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCF2COCF2Cl
CAS Number:
Molecular Weight:
198.93
MDL number:
UNSPSC Code:
12352103

vapor pressure

5.53 psi ( 20 °C)

assay

98%

refractive index

n20/D 1.33 (lit.)

bp

44-45 °C (lit.)

density

1.52 g/mL at 25 °C (lit.)

SMILES string

FC(F)(Cl)C(=O)C(F)(F)Cl

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service