Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D213756

Sigma-Aldrich

trans-p,p′-Diphenylstilbene

Synonym(s):

1,2-bis(p-Biphenylyl)ethylene, DPS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H20
CAS Number:
Molecular Weight:
332.44
EC Number:
MDL number:
Pricing and availability is not currently available.

SMILES string

c1ccc(cc1)-c2ccc(\C=C\c3ccc(cc3)-c4ccccc4)cc2

InChI

1S/C26H20/c1-3-7-23(8-4-1)25-17-13-21(14-18-25)11-12-22-15-19-26(20-16-22)24-9-5-2-6-10-24/h1-20H/b12-11+

InChI key

HXWQJYVUJPBQEW-VAWYXSNFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Suitable as a laser dye

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service