Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D209104

Sigma-Aldrich

Diphenyliodonium iodide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12I2
CAS Number:
Molecular Weight:
410.03
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

98%

SMILES string

[I-].[I+](c1ccccc1)c2ccccc2

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susana Castro et al.
Chemical communications (Cambridge, England), 48(72), 9089-9091 (2012-08-03)
A novel metal-free direct arylation of unbiased arenes with diaryliodonium triflates in a process where the use of solvents and additives is not required has been established.
Tao Ma et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 31(15), 5589-5595 (2011-04-15)
Generation of reactive oxygen species (ROS) causes cellular oxidative damage and has been implicated in the etiology of Alzheimer's disease (AD). In contrast, multiple lines of evidence indicate that ROS can normally modulate long-term potentiation (LTP), a cellular model for
T Franck et al.
Veterinary immunology and immunopathology, 130(3-4), 243-250 (2009-03-31)
Neutrophil (PMN) contribution to the acute inflammatory processes may lead to an excessive generation of reactive oxygen metabolites species (ROS) and secretion of granule enzymes. We compared the effects of either phorbol myristate acetate (PMA) or N-formyl-methionyl-leucyl-phenylalanine (fMLP) in combination
Liya Qin et al.
Journal of neuroinflammation, 9, 5-5 (2012-01-14)
Activation of microglia causes the production of proinflammatory factors and upregulation of NADPH oxidase (NOX) that form reactive oxygen species (ROS) that lead to neurodegeneration. Previously, we reported that 10 daily doses of ethanol treatment induced innate immune genes in
Sarah A Krawczyk et al.
PloS one, 7(4), e34904-e34904 (2012-04-12)
In obesity, there is an increase in reactive oxygen species (ROS) within adipose tissue caused by increases in inflammation and overnutrition. Hormone sensitive lipase (HSL) is part of the canonical lipolytic pathway and critical for complete lipolysis. This study hypothesizes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service