Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D138207

Sigma-Aldrich

2′,4′-Dimethylacetophenone

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3COCH3
CAS Number:
Molecular Weight:
148.20
Beilstein/REAXYS Number:
956772
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

96%

form

liquid

refractive index

n20/D 1.535 (lit.)

bp

117-118 °C/18 mmHg (lit.)

density

0.997 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1ccc(C)cc1C

InChI

1S/C10H12O/c1-7-4-5-10(9(3)11)8(2)6-7/h4-6H,1-3H3

InChI key

HSDSKVWQTONQBJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

212.0 °F - closed cup

flash_point_c

100.00 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masahiko Nakamura et al.
Bioorganic & medicinal chemistry, 17(13), 4740-4746 (2009-05-16)
Adult T cell leukemia (ATL), caused by infection of human T-lymphotropic virus type 1 (HTLV-1), has a poor prognosis and curative therapy is unavailable, so it is important to find or design superior lead compounds for the drug treatment of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service