D134600
Sinapinic acid
98%
Synonym(s):
3,5-Dimethoxy-4-hydroxycinnamic acid, 4-Hydroxy-3,5-dimethoxycinnamic acid, Sinapic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HOC6H2(OCH3)2CH=CHCO2H
CAS Number:
Molecular Weight:
224.21
Beilstein/REAXYS Number:
2699118
EC Number:
MDL number:
UNSPSC Code:
12352103
Recommended Products
assay
98%
mp
203-205 °C (dec.) (lit.)
SMILES string
COc1cc(\C=C\C(O)=O)cc(OC)c1O
replaced by
Product No.
Description
Pricing
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service