Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D126004

Sigma-Aldrich

Diisopropylfluorophosphate

Synonym(s):

DFP, DIFP, Diisopropyl phosphorofluoridate, Phosphoric acid diisopropyl ester fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CHO]2POF
CAS Number:
Molecular Weight:
184.15
Beilstein/REAXYS Number:
1723307
EC Number:
MDL number:
UNSPSC Code:
12352103

vapor pressure

0.58 mmHg ( 20 °C)

refractive index

n20/D 1.385 (lit.)

bp

62 °C/9 mmHg (lit.)

mp

−82 °C (lit.)

density

1.06 g/mL at 25 °C (lit.)

SMILES string

CC(C)OP(F)(=O)OC(C)C

InChI

1S/C6H14FO3P/c1-5(2)9-11(7,8)10-6(3)4/h5-6H,1-4H3

InChI key

MUCZHBLJLSDCSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Potent acetylcholinesterase inhibitor.[1][2]

Biochem/physiol Actions

Potent inhibitor of serine proteases and acetylcholinesterase; inhibits cathepsin G, cholinesterase, coagulation factor Xa, leucocyte elastase, pancreatic elastase, tissue kallikrein, plasmin, subtilisin, and thrombin. Inhibits apoptosis induced by ricin and bacterial toxins.
Potent inhibitor of serine proteases such as trypsin and chymotrypsin, and of acetylcholinesterase; also inhibits cathepsin G, cholinesterase, coagulation factor Xa, leucocyte elastase, pancreatic elastase, tissue kallikrein, plasmin, subtilisin, and thrombin. Inhibition of acetylcholinesterase makes this compound especially toxic. Inhibits apoptosis induced by ricin and bacterial toxins.

Analysis Note

Typically used at a concentration of 0.10 mM. A safer alternative inhibitor for serine protease is phenylmethylsulfonyl fluoride (PMSF).

replaced by

Product No.
Description
Pricing

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Marchot et al.
The Journal of biological chemistry, 268(17), 12458-12467 (1993-06-15)
Iodination of fasciculin 3 (FAS3) from Dendroaspis viridis venom provided us with a fully active specific probe of fasciculin binding sites on rat brain acetylcholinesterase (AChE). Binding and inhibition are concomitant, as association and inhibition rate constants k1 and ki
G Olmos et al.
European journal of pharmacology, 236(3), 467-476 (1993-06-04)
The specific binding of the agonists [3H]clonidine and [3H]UK 14304 (bromoxidine) and of the antagonist [3H]RX 821002 (2-metoxy idazoxan) to rat brain membranes, as well as clonidine-induced mydriasis, clonidine-induced inhibition of brain (3,4-dihydroxyphenylalaninme) DOPA synthesis and clonidine-induced inhibition of twitch
Supratik Dutta et al.
Bioorganic & medicinal chemistry, 18(6), 2265-2274 (2010-03-02)
Two new monocyclic analogs of the natural AChE inhibitor cyclophostin and two exocyclic enol phosphates were synthesized. The potencies and mechanisms of inhibition of the bicyclic and monocyclic enol phosphonates and the exocyclic enol phosphates toward human AChE are examined.
Gabriel Birkus et al.
Antimicrobial agents and chemotherapy, 51(2), 543-550 (2006-12-06)
GS-7340 and GS-9131 {9-[(R)-2-[[(S)-[[(S)-1-(isopropoxycarbonyl)ethyl]amino]phenoxyphosphinyl]methoxy]-propyl]adenine and 9-(R)-4'-(R)-[[[(S)-1-[(ethoxycarbonyl)ethyl]amino]phenoxyphosphinyl]methoxy]-2'-fluoro-1'-furanyladenine, respectively} are novel alkylalaninyl phenyl ester prodrugs of tenofovir {9-R-[(2-phosphonomethoxy)propyl]adenine} (TFV) and a cyclic nucleotide analog, GS-9148 (phosphonomethoxy-2'-fluoro-2', 3'-dideoxydidehydroadenosine), respectively. Both prodrugs exhibit potent antiretroviral activity against both wild-type and drug-resistant human immunodeficiency virus
Stefania Butini et al.
Journal of medicinal chemistry, 51(11), 3154-3170 (2008-05-16)
Protein conformational fluctuations are critical for biological functions, although the relationship between protein motion and function has yet to be fully explored. By a thorough bioinformatics analysis of cholinesterases (ChEs), we identified specific hot spots, responsible for protein fluctuations and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service