Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

D124702

Sigma-Aldrich

1,6-Diisocyanatohexane

98%

Synonym(s):

Hexamethylene diisocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OCN(CH2)6NCO
CAS Number:
Molecular Weight:
168.19
Beilstein/REAXYS Number:
956709
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

vapor pressure

0.05 mmHg ( 25 °C)

assay

98%

refractive index

n20/D 1.452 (lit.)

bp

255 °C (lit.)

density

1.04 g/mL at 25 °C (lit.)

SMILES string

O=C=NCCCCCCN=C=O

InChI

1S/C8H12N2O2/c11-7-9-5-3-1-2-4-6-10-8-12/h1-6H2

InChI key

RRAMGCGOFNQTLD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1C - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 1

flash_point_f

266.0 °F - Pensky-Martens closed cup

flash_point_c

130 °C - Pensky-Martens closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Bieler et al.
Occupational medicine (Oxford, England), 61(6), 440-442 (2011-08-10)
Occupational diisocyanate-induced extrinsic allergic alveolitis (EAA) is a rare and probably underestimated diagnosis. Two acute occupational EAA cases have been described in this context, but neither of them concerned hexamethylene diisocyanate (HDI) exposure. To investigate the cause of a life-threatening
Zibiao Li et al.
Biomacromolecules, 13(12), 3977-3989 (2012-11-22)
This paper reports the synthesis and characterization of new hyperbranched amphiphilic polyurethane multiblock copolymers consisting of poly(propylene glycol) (PPG), poly(ethylene glycol) (PEG), and polycaprolactone (PCL) segments as in situ thermogels. The hyperbranched poly(PPG/PEG/PCL urethane)s, termed as HBPEC copolymers, were synthesized
Sander M van Putten et al.
Journal of biomedical materials research. Part A, 98(4), 527-534 (2011-06-18)
Biomaterials are at continuous risk of bacterial contamination during production and application. In vivo, bacterial contamination of biomaterials delays the foreign body reaction (FBR). Endotoxins such as lipopolysaccharides (LPS), major constituents of the bacterial cell wall, are potent stimulators of
Xinbo Sun et al.
Acta biomaterialia, 8(4), 1498-1506 (2012-01-17)
An N-halamine precursor, 5,5-dimethylhydantoin (DMH), was covalently linked to the surface of polyurethane (PU) with 1,6-hexamethylene diisocyanate (HDI) as the coupling agent. The reaction pathways were investigated using propyl isocyanate (PI) as a model compound. The results suggested that the
Badri Sadat Jonaid et al.
Occupational and environmental medicine, 71(6), 415-422 (2014-04-01)
Isocyanate asthma is one of the most frequently identified forms of occupational asthma in industrialised countries. The underlying mechanisms have not been clarified. There is only limited information about the relationship between exhaled nitric oxide (eNO) and occupational exposure to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service