Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CF0039

Sigma-Aldrich

Urea crosslinker – C2-arm, NHS ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C13H14N4O9
CAS Number:
MDL number:
UNSPSC Code:
12352108

form

solid

reaction suitability

reagent type: cross-linking reagent

Application

The ureido-2,2´-diacetic acid bis(hydroxysuccinimide) ester, is the shortest version of MS-cleavable urea-based lysine-lysine reactive homobifunctional cross-linkers, featuring a C2-arm. At neutral or slightly basic pH, it irreversibly crosslinks the neighbouring lysine groups. The presence of the symmetrical urea moiety which is prone to collision-induced dissociation, allows for the unambiguous distinguishing of crosslinks in tandem CID-MS experiments.

Legal Information

Product of CF Plus Chemicals.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service