Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CDS025310

Sigma-Aldrich

3-[(tert-Butoxycarbonyl)amino]-5-hydroxybenzoic acid

AldrichCPR

Synonym(s):

3-[[(1,1-Dimethylethoxy)carbonyl]amino]-5-hydroxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C12H15O5N1
CAS Number:
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.21
Pricing and availability is not currently available.

SMILES string

N(c1cc(cc(c1)C(=O)O)O)C(=O)OC(C)(C)C

InChI

1S/C12H15NO5/c1-12(2,3)18-11(17)13-8-4-7(10(15)16)5-9(14)6-8/h4-6,14H,1-3H3,(H,13,17)(H,15,16)

InChI key

KSTCXQVSGVVIAO-UHFFFAOYSA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service