Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BLN00028

Sigma-Aldrich

3-Bromo-4-chloro-1H-pyrazolo[4,3-c]pyridine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$1,400.00

$1,400.00


Usually ships in 2 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 G
$1,400.00

About This Item

Empirical Formula (Hill Notation):
C6H3BrClN3
CAS Number:
Molecular Weight:
232.47
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

$1,400.00


Usually ships in 2 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

form

solid

SMILES string

Clc1nccc2[nH]nc(Br)c12

InChI

1S/C6H3BrClN3/c7-5-4-3(10-11-5)1-2-9-6(4)8/h1-2H,(H,10,11)

InChI key

CTZAGTLJGOBGEJ-UHFFFAOYSA-N

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service