Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

B1376

Sigma-Aldrich

5-Benzyloxy-6-methoxyindole

Synonym(s):

NSC 92529

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15NO2
CAS Number:
Molecular Weight:
253.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

powder

SMILES string

COc1cc2[nH]ccc2cc1OCc3ccccc3

InChI

1S/C16H15NO2/c1-18-15-10-14-13(7-8-17-14)9-16(15)19-11-12-5-3-2-4-6-12/h2-10,17H,11H2,1H3

InChI key

XDLZHWUPJHWVFK-UHFFFAOYSA-N

Application

Reactant for preparation of:
  • CB2 Cannabinoid receptor ligands
  • Potent antimitotic agents
  • Potent c-ABL inhibitors
  • Eumelanin pigment precursors

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service