Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ALD00380

Sigma-Aldrich

L-Fmoc-4-trifluoromethylphenylalanine

Synonym(s):

Fmoc-Phe(4-CF3)-OH, Fmoc-4-(trifluoromethyl)-L-phenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H20F3NO4
CAS Number:
Molecular Weight:
455.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

form

solid

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

color

beige

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

O=C(O)[C@@H](NC(OCC1C(C=CC=C2)=C2C3=C1C=CC=C3)=O)CC4=CC=C(C(F)(F)F)C=C4

InChI

1S/C25H20F3NO4/c26-25(27,28)16-11-9-15(10-12-16)13-22(23(30)31)29-24(32)33-14-21-19-7-3-1-5-17(19)18-6-2-4-8-20(18)21/h1-12,21-22H,13-14H2,(H,29,32)(H,30,31)/t22-/m0/s1

InChI key

YMEGJWTUWMVZPD-QFIPXVFZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Phenylalanine derivative was introduced in collaboration with Yu and coworkers in reflection to a methodology reported in C-H Activation.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Yu program focuses on efficient C–H bond activation for drug synthesis, using simple starting materials.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service