Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ALD00200

Sigma-Aldrich

2-Bromo-4,6-difluorobenzenesulfonyl fluoride

95% (GC)

Synonym(s):

2-Bromo-4,6-difluorobenzenesulfonyl fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2BrF3O2S
Molecular Weight:
275.04
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95% (GC)

form

liquid

reaction suitability

reaction type: click chemistry

refractive index

n/D 1.5133

density

1.8965 at 25 °C

SMILES string

FS(C1=C(Br)C=C(F)C=C1F)(=O)=O

InChI

1S/C6H2BrF3O2S/c7-4-1-3(8)2-5(9)6(4)13(10,11)12/h1-2H

InChI key

UYQJPKWCZMLUNP-UHFFFAOYSA-N

Application

Sulfonyl fluoride motif can be used as a connector for the assembly of -SO2- linked small molecules with proteins or nucleic acids. This new click chemistry approach through sulfates is a complimentary approach to using amides and phosphate groups as linkers.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service