Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ALD00074

Sigma-Aldrich

4-Fluoro-2-methylbenzenesulfonyl fluoride

95% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6F2O2S
CAS Number:
Molecular Weight:
192.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

95% (GC)

form

liquid

reaction suitability

reaction type: click chemistry

refractive index

n/D 1.4843

density

1.3918 at 25 °C

SMILES string

FC1=CC=C(S(=O)(F)=O)C=C1C

InChI

1S/C7H6F2O2S/c1-5-4-6(12(9,10)11)2-3-7(5)8/h2-4H,1H3

InChI key

LLHBXUKMIZJBMC-UHFFFAOYSA-N

Application

Sulfonyl fluoride motif can be used as a connector for the assembly of -SO2- linked small molecules with proteins or nucleic acids. This new click chemistry approach through sulfates is a complimentary approach to using amides and phosphate groups as linkers.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

206.0 °F

flash_point_c

96.67 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Sharpless Lab pursues useful new reactivity and general methods for selectively controlling chemical reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service