A65174
2-Amino-2-methyl-1,3-propanediol
99%
Synonym(s):
AMPD, Ammediol
About This Item
Recommended Products
vapor density
3.63 (vs air)
vapor pressure
~10 mmHg ( 152 °C)
assay
99%
useful pH range
7.8-9.7
pKa (25 °C)
8.8
bp
151 °C/10 mmHg (lit.)
mp
100-110 °C (lit.)
SMILES string
CC(N)(CO)CO
InChI
1S/C4H11NO2/c1-4(5,2-6)3-7/h6-7H,2-3,5H2,1H3
InChI key
UXFQFBNBSPQBJW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
replaced by
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
>212.0 °F
flash_point_c
> 100 °C
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service