Skip to Content
MilliporeSigma
All Photos(1)

Documents

A61004

Sigma-Aldrich

1-Methoxy-2-propylamine

95%

Synonym(s):

2-Amino-1-methoxypropane, 2-Methoxyisopropylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH(CH3)NH2
CAS Number:
Molecular Weight:
89.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

liquid

refractive index

n20/D 1.4065 (lit.)

bp

92.5-93.5 °C/743 mmHg (lit.)

density

0.845 g/mL at 25 °C (lit.)

SMILES string

COCC(C)N

InChI

1S/C4H11NO/c1-4(5)3-6-2/h4H,3,5H2,1-2H3

InChI key

NXMXETCTWNXSFG-UHFFFAOYSA-N

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

48.2 °F - closed cup

flash_point_c

9 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service