Skip to Content
MilliporeSigma
All Photos(1)

Documents

A45602

Sigma-Aldrich

2-Chloro-5-(trifluoromethyl)aniline

97%

Synonym(s):

3-Amino-4-chlorobenzotrifluoride, 6-Chloro-α,α,α-trifluoro-m-toluidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CF3)NH2
CAS Number:
Molecular Weight:
195.57
Beilstein/REAXYS Number:
879910
EC Number:
MDL number:
UNSPSC Code:
12352100

assay

97%

refractive index

n20/D 1.499 (lit.)

density

1.428 g/mL at 25 °C (lit.)

SMILES string

Nc1cc(ccc1Cl)C(F)(F)F

InChI

1S/C7H5ClF3N/c8-5-2-1-4(3-6(5)12)7(9,10)11/h1-3H,12H2

InChI key

VKTTYIXIDXWHKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service