Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

94902

Sigma-Aldrich

4′-Vinylbenzo-18-crown-6

technical, ≥90% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26O6
CAS Number:
Molecular Weight:
338.40
Beilstein/REAXYS Number:
1690412
MDL number:
UNSPSC Code:
12350000
PubChem Substance ID:

grade

technical

assay

≥90% (GC)

mp

56-60 °C

storage temp.

2-8°C

SMILES string

C=Cc1ccc2OCCOCCOCCOCCOCCOc2c1

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Monomer employed for the preparation of poly(18-crown-6) and related polymers[1][2][3]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A.J. Varma
Eur. Polymer J., 22, 111-111 (1986)
A. van Zon et al.
J. R. Neth. Chem. Soc., 100, 429-429 (1981)
J. Smid
Pure and Applied Chemistry. Chimie Pure Et Appliquee, 54, 2129-2129 (1982)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service