Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

94456

Sigma-Aldrich

Tributylmethylphosphonium methyl sulfate

≥95.0%

Synonym(s):

Methyltributylphosphonium methyl sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)3P(CH3)(CH3OSO3)
CAS Number:
Molecular Weight:
328.45
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥95.0%

form

liquid

density

1.027 g/mL at 20 °C (lit.)

SMILES string

COS([O-])(=O)=O.CCCC[P+](C)(CCCC)CCCC

InChI

1S/C13H30P.CH4O4S/c1-5-8-11-14(4,12-9-6-2)13-10-7-3;1-5-6(2,3)4/h5-13H2,1-4H3;1H3,(H,2,3,4)/q+1;/p-1

InChI key

JMXOUHNHEFFQIW-UHFFFAOYSA-M

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mauricio Llaver et al.
Talanta, 212, 120802-120802 (2020-03-03)
A simple, novel and highly sensitive ionic liquid-assisted cloud point extraction (IL-CPE) and preconcentration method was developed for the determination and speciation analysis of tellurium, using 1-octyl-3-methylimidazolium chloride as an additive for improving the extraction efficiency of Triton X-114. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service