Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

93977

Sigma-Aldrich

tert-Butyl 4-hydroxybutyrate

≥95.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2CH2COOC(CH3)3
CAS Number:
Molecular Weight:
160.21
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

assay

≥95.0% (GC)

form

liquid

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.431

density

0.976 g/mL at 20 °C

SMILES string

CC(C)(C)OC(=O)CCCO

InChI

1S/C8H16O3/c1-8(2,3)11-7(10)5-4-6-9/h9H,4-6H2,1-3H3

InChI key

RXGFTUPFXKYRMW-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

159.8 °F - closed cup

flash_point_c

71 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service