Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

938947

Sigma-Aldrich

4,6-Dichloropyrazolo[1,5-a]pyrazine

new

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3Cl2N3
CAS Number:
Molecular Weight:
188.01
MDL number:
UNSPSC Code:
12352142

Quality Level

assay

≥95%

form

powder or crystals

color

white to yellow

storage temp.

2-8°C

SMILES string

ClC1=CN2N=CC=C2C(Cl)=N1

InChI

InChI=1S/C6H3Cl2N3/c7-5-3-11-4(1-2-9-11)6(8)10-5/h1-3H

InChI key

ZSMFWODFQGCAKW-UHFFFAOYSA-N

General description

4,6-dichloropyrazolo[1,5-a]pyrazine is a halogenated heterocyclic building block useful in the synthesis of multiple pharmaceuticals and other valuable chemicals.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service