Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

937533

Sigma-Aldrich

4-[(Trifluoromethyl)sulfonyl]benzenesulfonyl chloride

≥95%

Synonym(s):

p-[(Trifluoromethyl)sulfonyl]benzenesulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4ClF3O4S2
CAS Number:
Molecular Weight:
308.68
MDL number:
UNSPSC Code:
12352142

Quality Level

assay

≥95%

form

powder or crystals

color

white to yellow

storage temp.

2-8°C

SMILES string

O=S(=O)(Cl)C1=CC=C(C=C1)S(=O)(=O)C(F)(F)F

InChI

InChI=1S/C7H4ClF3O4S2/c8-17(14,15)6-3-1-5(2-4-6)16(12,13)7(9,10)11/h1-4H

InChI key

PGNBXFXYMQFHLQ-UHFFFAOYSA-N

General description

4-[(Trifluoromethyl)sulfonyl]benzenesulfonyl chloride is a reagent used to introduce a trifluoromethylsulfonyl or other trifluoromethylated functional groups to a variety of organic compounds, including those of pharmaceutical, agrochemical, or material science interest.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service