Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92771

Sigma-Aldrich

3-(Trimethylsilyl)-2-oxazolidinone

≥97.0%

Synonym(s):

2-Oxo-3-(trimethylsilyl)-1,3-oxazolidine, TMSO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO2Si
CAS Number:
Molecular Weight:
159.26
Beilstein/REAXYS Number:
1210407
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥97.0%

form

liquid

refractive index

n20/D 1.455

bp

87-89 °C/0.8 mmHg

density

1.046 g/mL at 20 °C (lit.)

SMILES string

C[Si](C)(C)N1CCOC1=O

InChI

1S/C6H13NO2Si/c1-10(2,3)7-4-5-9-6(7)8/h4-5H2,1-3H3

InChI key

AUKCYOUETBBMFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Silylating agent, useful for the silylation of carboxylic acids, 1,3-dicarbonyl compounds and hydroxy compounds

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

176.0 °F - closed cup

flash_point_c

80 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lesbia Cristina Julio-Gonzalez et al.
Food research international (Ottawa, Ont.), 129, 108811-108811 (2020-02-11)
Enzymatic transgalactosylation, in different concentrated carbohydrate solutions, was investigated using brush border membrane vesicles (BBMV) from the pig small intestine. When lactulose was incubated with BBMV, the hydrolytic activity of the enzyme towards the disaccharide was observed to be very
Shubo Wang et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 7(5), 1903009-1903009 (2020-03-11)
As the power conversion efficiency (PCE) of perovskite solar cells (PSCs) is increased to as high over 25%, it becomes pre-eminent to study a scalable process with wide processing window to fabricate large-area uniform perovskite films with good light-trapping performance.
G.W. Perold
Journal of Chromatography A, 291, 365-365 (1984)
J.M. Aizpurua et al.
Canadian Journal of Chemistry, 62, 336-336 (1984)
Alvaro Ferreira-Lazarte et al.
Journal of agricultural and food chemistry, 67(16), 4662-4670 (2019-04-16)
Small intestinal brush border membrane vesicles from pig were used to digest galactooligosaccharides from lactose (GOS) and from lactulose (OsLu). Dissimilar hydrolysis rates were detected after digestion. Predominant glycosidic linkages and monomeric composition affected the resistance to intestinal digestive enzymes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service