Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

901820

Sigma-Aldrich

Invar electroplating solution

Synonym(s):

FeNi36 electroplating solution, Nickel-iron alloy electroplating solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352303
NACRES:
NA.23

form

liquid

contains

1 g/L±0.1 g/L stabilizer
0.01 g/L±0.001 g/L wetting agent
4 g/L±0.2 g/L Stress reducer

composition

9 g/L ±0.3 g/L NiBr2 · xH2O
24 g/L ±0.5 g/L H3BO3
51 g/L ±1.5 g/L FeSO4 · 7H2O
164 g/L ±5 g/L Ni(SO3NH2)2 .4H2O

Looking for similar products? Visit Product Comparison Guide

General description

Invar electroplating solution is designed for the fabrication of low stress, pinhole-free nickel-iron alloy films with extremely low coefficients of thermal expansion.

Features and Benefits

  • Deposits have low internal stress and are very ductile.
  • Near-zero linear thermal expansion in plated alloy films.
  • Surfaces are uniform and smooth with well-controlled thickness and excellent uniformity.
  • Qualified bath metrology for replenishment and a long shelf life.

Electroplated Invar film (5-10 μm thickness) properties:
1. Density: 8.1 g/cm3
2. Composition: 64 wt% Fe, 36 wt% Ni
3. Linear thermal expansion: 0.4 ± 0.1 * 10-6 K-1
4. Specific heat copacity at 573K: 0.45 ± 0.05 J/gK
5. Thermal conductivity: 43 ± 5 W/mK
6. Internal stress: 30 ± 2 MPa
7. Tensile strength: 500 ± 60 MPa
8. Yield strength: 280 ± 50 MPa
9. Ductility: 40 ±10 %

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 1A Inhalation - Eye Dam. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1 - STOT RE 2 Inhalation

target_organs

Respiratory Tract

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service