Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900982

Sigma-Aldrich

Menthide

≥98.0%

Synonym(s):

(4R,7S)-4-methyl-7-(1-methylethyl)oxepan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O2
CAS Number:
Molecular Weight:
170.25
EC Number:
UNSPSC Code:
12162002

assay

≥98.0%

form

liquid

color

colorless to pale yellow

storage temp.

−20°C

InChI

1S/C10H18O2/c1-7(2)9-5-4-8(3)6-10(11)12-9/h7-9H,4-6H2,1-3H3

InChI key

GGAXPLCKKANQED-UHFFFAOYSA-N

General description

Menthide is a seven-membered, plant-derived lactone monomer that can be readily polymerized via ring opening polymerization (ROP). Controlled ROP has yielded aliphatic polyesters with high molecular weights and narrow molecular weight distributions (PDI). Menthide has been copolymerized with other biologically derived monomers, such as lactide, to yield products such as thermoplastic elastomers and pressure-sensitive adhesives.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James A Wilson et al.
Biomacromolecules, 16(10), 3191-3200 (2015-09-18)
We report the one-pot copolymerization of ω-pentadecalactone (PDL) to produce tri- and diblock-like copolymers with the ability to undergo postpolymerization modification. The ε-substituted ε-lactone (εSL), menthide (MI), was copolymerized with PDL to introduce side chain functionality into poly(ω-pentadecalactone) (PPDL) copolymers.
Polyurethanes based on renewable polyols from bioderived lactones.
Gurusamy-Thangavelu SA, et al.
Polym. Chem., 3, 2941-2948 (2012)
Jihoon Shin et al.
Biomacromolecules, 13(11), 3833-3840 (2012-10-16)
Renewable ABA triblock copolymers were prepared by sequential polymerization of the plant-based monomers menthide and α-methylene-γ-butyrolactone (MBL or tulipalin A). Ring-opening transesterification polymerization of menthide using diethylene glycol as an initiator gave α,ω-dihydroxy poly(menthide) (HO-PM-OH), which was converted to α,ω-dibromo

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service