Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900968

Sigma-Aldrich

N,N′-Diphenyl-N,N′-bis-[4-(phenyl-m-tolylamino)phenyl]biphenyl-4,4′-diamine

greener alternative
Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C62H50N4
CAS Number:
Molecular Weight:
851.09
UNSPSC Code:
12352116
NACRES:
NA.23

assay

95%

form

solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Hole transport organic materials allow perfect energy level alignment with the absorber layer and therefore efficient charge collection, are prone to degradation in ambient conditions.Click here for more information.

Application

Hole transport material with ortho-linked terphenyl core structure for high power efficiency in blue phosphorescent org. light-emitting diodes.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A hole transport material with ortho-linked terphenyl core structure for high power efficiency in blue phosphorescent organic light-emitting diodes.
Lee CW and Lee JY
Organic Electronics, 15(2), 399-404 (2014)
Mounggon Kim et al.
Advanced materials (Deerfield Beach, Fla.), 27(15), 2515-2520 (2015-03-11)
High quantum efficiency above 18% and extended lifetime three times longer than that of phosphorescent organic light-emitting diodes (OLEDs) are demonstrated in blue thermally activated delayed fluorescent OLEDs.

Articles

Novel biosensing devices, like bio-FET sensors, offer label-free analysis by detecting ionic and biomolecular charges, crucial for diagnostics and biological research.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service