900818
1-Butyl-2,3-bis(dicyclohexylamino)cyclopropenimine
Synonym(s):
3-(Butylimino)-N1,N1,N2,N2-tetracyclohexylcycloprop-1-ene-1,2-diamine, n-Bu-C1-Cy HCl, CPI-Cy HCl
About This Item
Recommended Products
form
powder or crystals
reaction suitability
reagent type: catalyst
greener alternative product characteristics
Catalysis
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
greener alternative category
, Aligned
storage temp.
2-8°C
General description
Application
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Prof. Tristan Lambert and his co-coworkers have developed a new class of highly effective chiral Brønsted base catalysts, formulated from the unique cyclopropenimine architecture.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service