Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

900387

Sigma-Aldrich

Bis(9,9-dimethyl-9H-fluoren-7-yl)amine

Synonym(s):

N-(9,9-Dimethyl-9H-fluoren-2-yl)-9,9-dimethyl-9H-fluoren-2-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H27N
CAS Number:
Molecular Weight:
401.54
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

assay

>95%

form

powder or crystals

Storage temp.

2-8°C

InChI

1S/C30H27N/c1-29(2)25-11-7-5-9-21(25)23-15-13-19(17-27(23)29)31-20-14-16-24-22-10-6-8-12-26(22)30(3,4)28(24)18-20/h5-18,31H,1-4H3

Inchi Key

LCSMGMWMTSWXDD-UHFFFAOYSA-N

Application

Building block of organic sensitizers for use in dye-sensitized solar cells.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhancing the Performance of Organic Dye-Sensitized Solar Cells via a Slight Structure Modification.
Lim K, et al.
The Journal of Physical Chemistry C, 115(45), 22640-22646 (2011)
Wei-Shan Chao et al.
Chemical communications (Cambridge, England), 48(40), 4884-4886 (2012-04-14)
Hybrids based on a dibenzosuberene core bearing a spiro-fluorene junction at the C-5 position and with amino donor and β-thiophenyl-α-cyanoacrylic acid acceptor groups at C-3 and C-7, respectively, serve as new organic sensitizer materials for solar cell applications. Solar cell
Novel organic sensitizers containing a bulky spirobifluorene unit for solar cells.
Cho N, et al.
Tetrahedron, 65(31), 6236-6243 (2009)
Min Soo Kang et al.
Chemical communications (Cambridge, England), 48(75), 9349-9351 (2012-04-14)
A high solar-to-electricity conversion efficiency of 7.22% was achieved with a short circuit current (J(sc)) of 15.30 mA cm(-2), an open circuit voltage (V(oc)) of 669 mV and a fill factor (FF) of 0.71 for the 2Flu-ZnP-CN-COOH dye with a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service