Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

89155

Sigma-Aldrich

1-Ethyl-3-methylimidazolium tosylate

≥98.0% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$86.10
50 G
$667.00

$86.10

List Price$123.00Save 30%
Web-Only Promotion

Available to ship onApril 21, 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
$86.10
50 G
$667.00

About This Item

Empirical Formula (Hill Notation):
C13H18N2O3S
CAS Number:
Molecular Weight:
282.36
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$86.10

List Price$123.00Save 30%
Web-Only Promotion

Available to ship onApril 21, 2025Details


Request a Bulk Order

Quality Level

assay

≥98.0% (HPLC)

form

liquid

impurities

≤1.0% water

refractive index

n20/D 1.548

mp

25-35 °C

density

1.231 g/mL at 20 °C (lit.)

functional group

tosylate

SMILES string

CCn1cc[n+](C)c1.Cc2ccc(cc2)S([O-])(=O)=O

InChI

1S/C7H8O3S.C6H11N2/c1-6-2-4-7(5-3-6)11(8,9)10;1-3-8-5-4-7(2)6-8/h2-5H,1H3,(H,8,9,10);4-6H,3H2,1-2H3/q;+1/p-1

InChI key

HXMUPILCYSJMLQ-UHFFFAOYSA-M

Physical form

non halogenated ionic liquid

Other Notes

Ionic liquid solvent for aromatic substitutions[1][2]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K.K. Laali, V.J. Gettwert
Journal of Fluorine Chemistry, 107, 31-31 (2001)
K K Laali et al.
The Journal of organic chemistry, 66(1), 35-40 (2001-06-30)
Potential utility of a series of 1-ethyl-3-methylimidazolium salts [emim][X] with X = OTf-, CF3COO-, and NO3- as well as [HNEtPri2][CF3COO] (protonated Hünig's base) ionic liquids were explored as solvent for electrophilic nitration of aromatics using a variety of nitrating systems

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service