Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

86633

Sigma-Aldrich

Tetraethylammonium hydroxide solution

~40% in H2O

Synonym(s):

TEAOH solution, TEA hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)4N(OH)
CAS Number:
Molecular Weight:
147.26
Beilstein/REAXYS Number:
3912961
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

~40% in H2O

impurities

≤0.1% halides (as chloride)

refractive index

n20/D 1.422

anion traces

sulfate (SO42-): ≤1000 mg/kg

functional group

amine

SMILES string

[OH-].CC[N+](CC)(CC)CC

InChI

1S/C8H20N.H2O/c1-5-9(6-2,7-3)8-4;/h5-8H2,1-4H3;1H2/q+1;/p-1

InChI key

LRGJRHZIDJQFCL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Tetraethylammonium hydroxide solution is widely used as a templating agent / structure-directing agent in the synthesis of ferric oxide nanorods, nanocrystalline yttrium oxide powder, silicoaluminophosphate (SAPO) molecular sieves and zeolites.[1][2][3][4]

Caution

may form precipitate on storage, can be redissolved on heating to 40°C

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of porous α-Fe2O3 nanorods and deposition of very small gold particles in the pores for catalytic oxidation of CO.
Zhong, Ziyi et al.
Chemistry of Materials, 19(19), 4776-4782 (2007)
Static hydrothermal crystallization of SUZ-4 zeolite in the presence of seed and tetraethylammonium hydroxide.
Zhou, Hualan and Wu, Yajing and Zhang, Wei and Wang, Jun
Materials Chemistry and Physics, 134(2-3), 651-656 (2012)
Synthesis and characterization of nanocrystalline yttrium oxide prepared with tetraalkylammonium hydroxides.
Fokema, Mark D et al.
Langmuir, 16(7), 3154-3159 (2000)
Methanol conversion on SAPO-34 catalysts prepared by mixed template method.
Lee, Yun-Jo et al.
Applied Catalysis A: General, 329(19), 130-136 (2007)
Min Shen et al.
Vascular pharmacology, 58(1-2), 64-70 (2012-07-24)
Evidence has indicated that resveratrol (Res) produces vasorelaxation and may decrease the coronary heart disease mortality. However, several pathways involved in the mechanism of vasorelaxation are still unclear. This study was designed, therefore, to test the probable ion channels or

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service