Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

861499

Sigma-Aldrich

S-(4-Nitrobenzyl)-6-thioinosine

98%

Synonym(s):

6-[(4-Nitrobenzyl)thio]-9-β-D-ribofuranosylpurine, NBMPR, NBTI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17N5O6S
CAS Number:
Molecular Weight:
419.41
EC Number:
MDL number:
UNSPSC Code:
12352200

assay

98%

mp

187-190 °C (lit.)

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(SCc4ccc(cc4)[N+]([O-])=O)ncnc23

InChI

1S/C17H17N5O6S/c23-5-11-13(24)14(25)17(28-11)21-8-20-12-15(21)18-7-19-16(12)29-6-9-1-3-10(4-2-9)22(26)27/h1-4,7-8,11,13-14,17,23-25H,5-6H2/t11-,13-,14-,17-/m1/s1

Inchi Key

DYCJFJRCWPVDHY-LSCFUAHRSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A very potent inhibitor of nucleoside transport. NBMPR, in cultures, binds tightly but reversibly to tumor membrane sites associated with the nucleoside transport mechanism and is, therefore, a useful tool for cancer research.

Biochem/physiol Actions

Inhibitor of equilibrative nucleoside transporters (ENTs), particularly adenosine transporters, in central nervous system and vascular smooth muscle.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service