Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

808601

Sigma-Aldrich

(Bromodifluoromethyl)trimethylsilane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9BrF2Si
CAS Number:
Molecular Weight:
203.10
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

description

Flash point: 46.4°C

assay

98%

form

liquid

refractive index

n/D 1.407

density

1.306

storage temp.

−20°C

SMILES string

C[Si](C)(C(F)(F)Br)C

InChI

1S/C4H9BrF2Si/c1-8(2,3)4(5,6)7/h1-3H3

InChI key

WDZVWBWAUSUTTO-UHFFFAOYSA-N

General description

(Bromodifluoromethyl)trimethylsilane (TMSCF2Br) is commonly used as a source to generate dilfluorocarbene.[1][2] TMSCF3 (Ruppert–Prakash reagent) and BBr3 undergoes fast halogen–exchange reaction to form MSCF2Br.[3]

Application

(Bromodifluoromethyl)trimethylsilane may be used in the preparation of:
  • gem-difluorocyclopropa(e)nes[3]
  • O-, S-, N-, and P-difluoromethylated compounds[3]
  • (chlorodifluoromethyl)trimethylsilane[4]
As reported by Hu and co-workers, (Bromodifluoromethyl)trimethylsilane is a highly useful reagent for the formation of difluoromethene-containing 3-membered rings and can also difluoromethylate heteroatoms with the assistance of alkaline bases, most effectively KOH.

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

115.5 °F

flash_point_c

46.4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent advances in the synthetic application of difluorocarbene.
Ni C and Hu J.
Synthesis, 46(07), 842-863 (2014)
Nucleophilic Bromodifluoromethylation of Iminium Ions.
Tsymbal AV, et al.
The Journal of Organic Chemistry, 79(17), 7831-7835 (2014)
Synthesis of gem-Difluorocyclopropa (e) nes and O-, S-, N-, and P-Difluoromethylated Compounds with TMSCF2Br.
Li L, et al.
Angewandte Chemie (International Edition in English), 52(47), 12390-12394 (2013)
Coupling of a,a-difluoro-substituted organozinc reagents with 1-bromoalkynes.
Zemtsov AA, et al.
Beilstein Journal of Organic Chemistry, 11(1), 2145-2149 (2015)

Related Content

Prof. Jinbo Hu's lab focuses on developing new fluorination reagents and reactions, including difluoromethylation and monofluoromethylation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service