Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

805289

Sigma-Aldrich

2-fluoro-4-methylphenylboronic acid MIDA ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13BFNO4
Molecular Weight:
265.05
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

95% (HPLC)

form

powder

mp

188.4 °C

functional group

fluoro

SMILES string

O=C(CN(C)C1)OB(C2=C(F)C=C(C)C=C2)OC1=O

InChI

1S/C12H13BFNO4/c1-8-3-4-9(10(14)5-8)13-18-11(16)6-15(2)7-12(17)19-13/h3-5H,6-7H2,1-2H3

InChI key

NAIYXIBAIMFIER-UHFFFAOYSA-N

General description

2-Fluoro-4-methylphenylboronic acid MIDA ester, an aryl MIDA (N-methyliminodiacetic acid) boronate, is a MIDA-protected boronate ester. It participates in the Suzuki-Miyaura cross-coupling reactions.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service