Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

805262

Sigma-Aldrich

2-fluoro-3-methoxyphenylboronic acid MIDA ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13BFNO5
Molecular Weight:
281.04
UNSPSC Code:
12352103
PubChem Substance ID:

form

powder

mp

223 °C

SMILES string

O=C(CN(C)C1)OB(C2=C(F)C(OC)=CC=C2)OC1=O

InChI

1S/C12H13BFNO5/c1-15-6-10(16)19-13(20-11(17)7-15)8-4-3-5-9(18-2)12(8)14/h3-5H,6-7H2,1-2H3

InChI key

DEOGQIAPGXNRBR-UHFFFAOYSA-N

General description

2-Fluoro-3-methoxyphenylboronic acid MIDA ester, an aryl MIDA (N-methyliminodiacetic acid) boronate, is a MIDA-protected boronate ester. It participates in Suzuki-Miyaura cross-coupling reactions.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service