Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

803006

Sigma-Aldrich

Acrylonitrile-13C3,d3

≥99 atom % 13C, ≥98 atom % D, ≥98% (CP), contains hydroquinone as stabilizer

Synonym(s):

2-Propenenitrile-13C3,d3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
D213C=13CD13CN
Molecular Weight:
59.06
UNSPSC Code:
12142201
PubChem Substance ID:

isotopic purity

≥99 atom % 13C
≥98 atom % D

assay

≥98% (CP)

form

liquid

contains

hydroquinone as stabilizer

mass shift

M+6

storage temp.

2-8°C

SMILES string

N#C[13C]([2H])=[13C]([2H])[2H]

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1B - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

23.0 °F - closed cup

flash_point_c

-5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service