Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

802581

Sigma-Aldrich

Aquivion® PW79S-Li

coarse powder, PFSA eq. wt. 790 g/mole SO3Li, stabilized CF3 polymer chain ends

Synonym(s):

Aquivion® SO3Li-form, Tetrafluoroethylene-perfluoro(3-oxa-4-pentenesulfonic acid) copolymer, Ethanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
26111700

description

eq wt.: 790 g/mole

form

coarse powder
powder

Application

Aquivion® PW79S-Li can be used, amongst other, in chemical or electrochemical structures or devices where Lithium ion conductivity of a solid conductor is requested. The powder can be mechanically processed into blends, films or coated on substrates, preferably accompanied or followed by a heat treatment. For casting processes Aquivion® D79-25BS-Li dispersion is a more recommendable ingredient.

Preparation Note

  • Keep away from heat and sources of ignition.
  • Avoid thermal decomposition caused by overheating the resin.
  • Take measures to prevent build-up of electrostatic charge.

Legal Information

Aquivion is a registered trademark of Syensqo Group

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Advances in electrochemical water conversion and understanding PEMFC degradation drive progress in hydrogen technologies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service