Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

797405

Sigma-Aldrich

6-Benzhydryl-3,3-difluoro-1,6-diazaspiro[3.3]heptane hydrochloride salt

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19ClF2N2
Molecular Weight:
336.81
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

powder

functional group

fluoro
phenyl

storage temp.

2-8°C

SMILES string

FC1(F)CNC21CN(C(C3=CC=CC=C3)C4=CC=CC=C4)C2

InChI

1S/C18H18F2N2/c19-18(20)11-21-17(18)12-22(13-17)16(14-7-3-1-4-8-14)15-9-5-2-6-10-15/h1-10,16,21H,11-13H2

InChI key

GQOMJRFLSOTZSO-UHFFFAOYSA-N

Application

Spirocyclic building block is part of a series of advanced angular[3.3]heptanes developed by Carreira and coworkers.

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Carreira Research & SpiroChem innovate spirocyclic building blocks, enhancing drug discovery with diverse functionalization sites and compact modules.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service