Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

792403

Sigma-Aldrich

5,5′-Dibromo-4,4′-ditetradecyl-2,2′-bithiophene

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H60Br2S2
Molecular Weight:
716.80
UNSPSC Code:
12352103
PubChem Substance ID:

assay

96%

form

powder

mp

64-68 °C

SMILES string

BrC1=C(CCCCCCCCCCCCCC)C=C(C2=CC(CCCCCCCCCCCCCC)=C(Br)S2)S1

InChI

1S/C36H60Br2S2/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-31-29-33(39-35(31)37)34-30-32(36(38)40-34)28-26-24-22-20-18-16-14-12-10-8-6-4-2/h29-30H,3-28H2,1-2H3

InChI key

YWKJVWGQFAPEDZ-UHFFFAOYSA-N

Application

This monomer is used in the synthesis of semiconductive polymers for OPV and OFET devices

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Itaru Osaka et al.
Journal of the American Chemical Society, 132(14), 5000-5001 (2010-03-20)
We have designed and synthesized novel semiconducting polymers by introducing naphtho[1,2-b:5,6-b']dithiophene (NDT) into the polythiophene backbone. These polymers, which have a highly pi-extended heteroarene unit, achieved mobilities (>0.5 cm(2) V(-1) s(-1)) that are among the highest recorded to date for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service