Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

791989

Sigma-Aldrich

PDPP2T-TT-OD

Synonym(s):

Diketopyrrolopyrrole - thienothiophene copolymer, Diketopyrrolopyrrole p-type semiconductor, Xerox XSC4p

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C60H90N2O2S4)n
CAS Number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

solid

mol wt

average Mw 40,000-60,000 by GPC

color

dark blue

mp

>200 °C

λmax

820 nm (thin film)

orbital energy

HOMO 5.2 eV 

OFET device performance

Bottom gate top contact device with silane modified SiO2 dielectric.Processing method spin coating; thermal annealing at 140 °C/ 10 min

  • Semiconductor Type:
  • Mobility: 0.60 cm2/V·s
  • On/Off Ratio: 10^6 - 10^7

PDI

2.5‑3.0

General description

PDPP2T-TT-OD is thiophene based conducting polymer with diketopyrrolo-pyrrole (DPP) as the electron deficient group and octyldodecyl acting as a pendant attachment. Its structure has a DPP-thiophene and a variable bithiophene (2T). It has a high charge carrying mobility that makes it useful in the development of organic electronics based devices.

Application

PDPP2T-TT-OD can be used as a π-conjugating polymer that can be used in the fabrication of a variety of devices such as organic thin film transistors (OTFTs), photovoltaic cells, polymeric solar cells and field effect transistors (FETs).
Xerox demonstrated device performance on a n-doped silicon wafer with 100 - 300 nm native SiO2 dielectric modified with an octadecyltrichlorosilane self-assembled
monolayer. An XSC4p film was deposited by spin-coating a 0.5 – 0.7 wt. % polymer solution at 1000 rpm for 60 sec. The film was dried in a vacuum oven for 10 min at 80
°C and then thermally annealed at 140 °C for 10 min. Gold source-drain electrodes were deposited by vacuum evaporation using a shadow mask (90 micrometer channels).
The electrical performance was characterized using a Keithley SCS-4200 system. The reported mobility is the average saturated mobility of four devicεs ± the standard
deviation.

Legal Information

Product of Xerox®. Xerox® is a registered trademark of Xerox Corporation.
Xerox is a registered trademark of Xerox Corporation

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weiwei Li et al.
Advanced materials (Deerfield Beach, Fla.), 25(23), 3182-3186 (2013-03-07)
A high-molecular-weight conjugated polymer based on alternating electron-rich and electron-deficient fused ring systems provides efficient polymer solar cells when blended with C60 and C70 fullerene derivatives. The morphology of the new polymer/fullerene blend reduces bimolecular recombination and allows reaching high
Molecular packing of high-mobility diketo pyrrolo-pyrrole polymer semiconductors with branched alkyl side chains
Zhang X, et al.
Journal of the American Chemical Society, 133(38), 15073-15084 (2011)
Xinran Zhang et al.
Journal of the American Chemical Society, 133(38), 15073-15084 (2011-08-06)
We describe a series of highly soluble diketo pyrrolo-pyrrole (DPP)-bithiophene copolymers exhibiting field effect hole mobilities up to 0.74 cm(2) V(-1) s(-1), with a common synthetic motif of bulky 2-octyldodecyl side groups on the conjugated backbone. Spectroscopy, diffraction, and microscopy
Design of high-mobility diketopyrrolopyrrole-based pi-conjugated copolymers for organic thin-film transistors
Yi Z, et al.
Advanced Materials, 27(24), 3589-3606 (2015)
High mobility thiazole-diketopyrrolopyrrole copolymer semiconductors for high performance field-effect transistors and photovoltaic devices
Subramaniyan S, et al.
Macromolecules, 45(22), 9029-9037 (2012)

Articles

The development of high-performance conjugated organic molecules and polymers has received widespread attention in industrial and academic research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service