Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

777579

Sigma-Aldrich

1,1-Difluoro-2-vinylcyclopropane

97%

Synonym(s):

2-Ethenyl-1,1-difluorocyclopropane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6F2
CAS Number:
Molecular Weight:
104.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.368 (lit.)

density

0.986 g/mL at 25 °C

functional group

fluoro

storage temp.

2-8°C

SMILES string

FC1(F)CC1C=C

InChI

1S/C5H6F2/c1-2-4-3-5(4,6)7/h2,4H,1,3H2

InChI key

QCLGRUGHFNFAFX-UHFFFAOYSA-N

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

71.7 °F

flash_point_c

22.05 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service