Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

776424

Sigma-Aldrich

Poly(tert-butyl acrylate), DDMAT terminated, azide terminated

average Mn 8,500, PDI ≤1.2

Synonym(s):

poly(t-butyl acrylate) RAFT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352100
NACRES:
NA.23

form

solid

mol wt

average Mn 8,500

transition temp

Tm 241-246 °C

PDI

≤1.2

Application

Poly(tert-butyl acrylate) is a hydrophobic polymer used in many applications; primarily in which the tert-butyl group is substituted with a desired functionality. The hydrophilicity of this block can be tuned by varying the hydrolysis of the t-butyl group. The end group functionality contains an azide as well as the 2-(Dodecylthiocarbonothioylthio)-2-methylpropionic acid RAFT agent moiety. This RAFT CTA will allow for additional polymerization to generate a diblock copolymer. This RAFT agent is especially suited for the polymerization of styrene; acrylate and acrylamide monomers.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Reversible addition–fragmentation chain transfer (RAFT) polymerization is rapidly moving to the forefront in construction of drug and gene delivery vehicles.

Over the past two decades, the rapid advance of controlled living polymerization (CLP) techniques.

Microparticles in drug delivery: Study on controlling chitosan microparticle size and distribution, exploring encapsulation of BSA and TPP cross-linker.

The modification of biomacromolecules, such as peptides and proteins, through the attachment of synthetic polymers has led to a new family of highly advanced biomaterials with enhanced properties.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service