Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

775592

Sigma-Aldrich

5-Bromo-2-chloro-3-methoxypyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5BrClNO
CAS Number:
Molecular Weight:
222.47
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

mp

68-72 °C

SMILES string

BrC1=CC(OC)=C(Cl)N=C1

InChI

1S/C6H5BrClNO/c1-10-5-2-4(7)3-9-6(5)8/h2-3H,1H3

InChI key

GFJMOZYIDADKLJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Skull and crossbonesCorrosion

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service