Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

769746

Sigma-Aldrich

Methyl 3-hydroxybutyrate

≥95.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2COOCH3
CAS Number:
Molecular Weight:
118.13
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

assay

≥95.0% (GC)

form

liquid

SMILES string

O=C(OC)CC(O)C

InChI

1S/C5H10O3/c1-4(6)3-5(7)8-2/h4,6H,3H2,1-2H3

InChI key

LDLDJEAVRNAEBW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

remarks on GC: no single impurity > 1.0%

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

321.8 °F

flash_point_c

161 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service