Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

766313

Sigma-Aldrich

1H-Benzotriazole-1-sulfonyl azide

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4N6O2S
CAS Number:
Molecular Weight:
224.20
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

solid

mp

80-90 °C

storage temp.

−20°C

SMILES string

[N-]=[N+]=NS(=O)(=O)n1nnc2ccccc12

InChI

1S/C6H4N6O2S/c7-9-11-15(13,14)12-6-4-2-1-3-5(6)8-10-12/h1-4H

InChI key

AYOJLIHUPZVAHM-UHFFFAOYSA-N

Application

A crystalline and stable diazotransfer reagent to provide azides (from amines) and diazo species.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alan R Katritzky et al.
The Journal of organic chemistry, 75(19), 6532-6539 (2010-09-10)
Benzotriazol-1-yl-sulfonyl azide, a new crystalline, stable, and easily available diazotransfer reagent provides N-(α-azidoacyl)benzotriazoles convenient for N-, O-, C- and S-acylations. The efficient syntheses of various amides, azido protected peptides, esters, ketones and thioesters is reported together with a wide range

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service