Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

760684

Sigma-Aldrich

Methyl 4-bromo-2-chloropyridine-3-carboxylate

97%

Synonym(s):

Methyl 4-bromo-2-chloronicotinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5BrClNO2
CAS Number:
Molecular Weight:
250.48
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

54-58 °C

SMILES string

COC(=O)c1c(Cl)nccc1Br

InChI

1S/C7H5BrClNO2/c1-12-7(11)5-4(8)2-3-10-6(5)9/h2-3H,1H3

InChI key

FITBMOURWDPXGV-UHFFFAOYSA-N

pictograms

Environment

signalword

Warning

hcodes

pcodes

Hazard Classifications

Aquatic Acute 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christoph Gibson et al.
Journal of medicinal chemistry, 52(14), 4370-4379 (2009-06-26)
Blockade of the bradykinin B(2) receptor provides therapeutic benefit in hereditary angioedema (HAE) and potentially in many other diseases. Herein, we describe the development of highly potent B(2) receptor antagonists with a molecular weight of approximately 500 g/mol. First, known

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service