Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

760412

Sigma-Aldrich

Niobium(V) ethoxide

packaged for use in deposition systems

Synonym(s):

NbOEt, Niobium ethanolate, Niobium ethylate, Niobium pentaethoxide, Pentaethoxyniobium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Nb(OCH2CH3)5
CAS Number:
Molecular Weight:
318.21
Beilstein/REAXYS Number:
3678883
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

96.5-103.5% (gravimetric)
99.999% (trace metals analysis)

form

liquid

reaction suitability

core: niobium

refractive index

n20/D 1.516 (lit.)

bp

142 °C/0.1 mmHg (lit.)

mp

5-6 °C (lit.)

density

1.268 g/mL at 25 °C (lit.)

SMILES string

CCO[Nb](OCC)(OCC)(OCC)OCC

InChI

1S/5C2H5O.Nb/c5*1-2-3;/h5*2H2,1H3;/q5*-1;+5

InChI key

ZTILUDNICMILKJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Niobium(V) ethoxide is a colorless liquid that is miscible with some organic solvents, but which hydrolyzes readily. Niobium(V) ethoxide boils at 142 °C at 0.1 mmHg.

Application

Niobium ethoxide is an important precursor for depositing thin films of niobium oxide and other thin films doped with niobium.[1] Because of its low boiling point and low decomposition temperature (~325-350 °C), niobium(V) ethoxide is used as a precursor in atomic-layer deposition (ALD)[2], chemical vapor deposition (CVD)[3], and metal-organic chemical vapor deposition (MOCVD)[4] of niobium oxides.

Features and Benefits

Niobium(V)ethoxide packaged for use in deposition systems is packaged in a Swagelokstainless-steel deposition system convenient for connecting to ALD, CVD, andMOCVD systems.

  • Steel cylinder connected to 316stainless steel ball-valve
  • 1/4 inch male Swagelok VCRconnections

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

96.8 °F - closed cup

flash_point_c

36 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biqiong Wang et al.
ACS applied materials & interfaces, 10(2), 1654-1661 (2017-12-09)
The development of solid-state electrolytes by atomic layer deposition (ALD) holds unparalleled advantages toward the fabrication of next-generation solid-state batteries. Lithium niobium oxide (LNO) thin films with well-controlled film thickness and composition were successfully deposited by ALD at a deposition
Huang, Y.; et al.
Applied Surface Science, 257, 7305-7305 (2011)
Epitaxial Pb(Mg1/3Nb2/3)O3 thin films synthesized by metal-organic chemical vapor deposition.
Bai G R, et al.
Applied Physics Letters, 76, 3106 -3106 (2000)
Sanjayan Sathasivam et al.
ACS applied materials & interfaces, 9(21), 18031-18038 (2017-05-12)
Nb2O5 is an important material able to exist in many polymorphs with unique optical properties and morphologies that are dependent on the synthetic route. Here we report a novel ambient-pressure chemical vapor deposition route to Nb2O5 via aerosol-assisted chemical vapor
Yujing Liu et al.
ACS nano, 4(9), 5373-5381 (2010-08-26)
Crystalline niobium-doped titania nanoparticles were synthesized via solvothermal procedures using tert-butyl alcohol as a novel reaction medium, and their assembly into mesoporous films was investigated. The solvothermal procedure enables the preparation of crystalline doped and undoped nonagglomerated titania nanoparticles, whose

Articles

The properties of many devices are limited by the intrinsic properties of the materials that compose them.

Questions

  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service